SP Wilmad-LabGlass High Pressure Vacuum NMR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
This pressure/vacuum tube is a highly reliable NMR tube for medium range pressure (<300 psi) experiments.
- PV-ANV valve is made of PTFE and all other parts are borosilicate glass
- Valve is opened simply by turning counterclockwise
Designed to connect to a ¹/₈" metal (stainless steel or brass) vacuum line using SwageLok® fittings or a rubber vacuum hose and a glass connector.
Customers Who Bought This Also Bought