Spécifications
- Sterility:Non sterile
- Description:3-ply ear loop facemask
- Cat. no.:470329-608
Spécifications
A propos de cet article
Facemask offers a Bacterial Filtration Efficiency (BFE) of <95%.
- Three layers of protection
- Non sterile
- Manufactured in an FDA registered facility
Safe, comfortable, and easy to use. This 3-ply ear loop facemask features three layers of protection, soft ear loops, and an adjustable nose cover. They are disposable, flat pleated, and easy to put on and remove.