Disposable 3-Ply Facemasks
# de catalogue 470329-602
Fournisseur: Wards
Spécifications
- Description:Disposable facemasks
- Cat. no.:470329-602
Spécifications
A propos de cet article
Non-surgical facemasks.
- Breathable
- Stretchy, elastic ear loops
- Excellent for general-purpose, everyday use
Disposable, non-surgical 3-ply face masks feature a melt-blown polypropylene filtration layer. Masks include three, non-woven layers for optimal comfort and a filter for added protection. It is recommended to replace masks every 3 to 5 hours. Always wear with the blue-side facing out and the nose strip at the top.
Individual masks measure 3.6×6.75".
Packaging: Masks are packaged in a box of 50.
Customers Who Bought This Also Bought