Avantor® ACE®, Replacement Ferrules
# de catalogue 76408-450
Fournisseur: HICHROM LIMITED
Spécifications
- Description:Replacement PEEK ferrules for UHPLC guard cartridge holder for 2,1 to 4,6 mm ID columns
- Cat. no.:76408-450