ContactUsLinkComponent
Chromogenic Analysis of Water Contaminants
Chromogenic Analysis of Water Contaminants
# de catalogue 470107-598
Fournisseur:  Edvotek
EDVOTEK WATER QUALITY TESTING 1 KIT
Chromogenic Analysis of Water Contaminants
# de catalogue 470107-598
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Chromogenic Analysis of Water Contaminants
  • No. of Lab Groups:
    10
  • Cat. no.:
    470107-598
  • Supplier No.:
    951
Spécifications
A propos de cet article
Test water from your own environment for fecal coliforms.
  • Complete assay in 30 minutes, and grow overnight
  • Non-pathogenic bacteria provided
  • Easy to interpret results
  • Includes instructor's guide and discussion questions
In this experiment, students will test control and environmental water samples for the presence of total coliform bacteria and E. coli using color changing enzyme assays. Testing drinking water for every possible type of pathogenic bacteria is slow and costly. Thus, drinking water is tested for coliforms - including the familiar E. coli. Presence of coliforms is an indicator of fecal contamination.
Bactobead refill contains positive control organisms: Escherichia coli, and Citrobacter freundii.
BodyTextAccessories: Additional equipment needed but not included: Long wave UV Lamp, 10% bleach solution or other disinfectant, water-collection bottles (e.g. screw-cap drink bottles). Water bath or incubator and micropipettes are helpful but not required.
Ordering information: Items will ship immediately upon ordering. Please call to place your order if you would like to specify a future date for shipment.
Delivery: Some components require Refrigerated storage upon receipt.
Customers Who Bought This Also Bought