SP Wilmad-LabGlass Septa for Screw-Cap NMR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Replacement septa for NMR screw-cap tubes.
- Made of PTFE/rubber or PTFE/silicone
PTFE/silicone septums are inert to most organic solvents and compounds but not recommended for strongly corrosive materials. The septum remains reliable after multiple punctures.
Customers Who Bought This Also Bought