SP Wilmad-LabGlass Sapphire NMR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Sapphire NMR tubes have extraordinary hardness and a high melting temperature around 2000 °C making them ideal for high pressure experiments up to 5801.5 psi.
- Outstanding chemical resistance to corrosive solvents, including HF acid
- Provides zero background in most NMR experiments
- Guaranteed straightness for optimized shimming quality and tested in spinner turbines
Saphire NMR tubes are popular among the following applications: High Pressure NMR up to 5801.5 psi, tube only, no flange supplied; 19F studies, especially studies with HF acid involved; 29Si studies.
Caution: Epoxy glue is recommended if connecting to a metal gas line.
Customers Who Bought This Also Bought