SP Wilmad-LabGlass Precision L, S, and X Band EPR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Precision EPR tubes have 10 to 15 times tighter tolerances than standard tubes.
- Available in Quartz and Suprasil
- Individually inspected for impurities and quality
- Includes disposable grade cap
Ideal for experiments that require a constant filing factor or reproducibility on sample volume in the cavity.
Customers Who Bought This Also Bought