SP Wilmad-LabGlass Economy L, S, and X Band EPR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Ideal for high throughput EPR L, S, and X band EPR experiments that do not require tight camber or concentricity tolerances.
- Manufactured from High-quality Quartz and Suprasil
- Economically priced
- Includes disposable grade cap
They are individually inspected for impurities and quality.
Customers Who Bought This Also Bought