SP Wilmad-LabGlass X Band Capillary Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Wilmad's X-band EPR tubes are made from clear fused quartz with the lowest impurities to guarantee success in detecting subtle spectroscopic details.
- Available with both ends open
- Ideal for applications with limited sample size
- Easy to seal with a hot flame or sealant of your choice
Rigorous testing is done on each tube.
Customers Who Bought This Also Bought