SP Wilmad-LabGlass Q Band Standard EPR Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Wilmad's Q-band EPR tubes are manufactured of high quality materials to give optimal performance in these high frequency applications.
- Available with both ends open or sealed at one end
- Improved SNR
Rigorous testing is done on each tube.
Customers Who Bought This Also Bought