SP Wilmad-LabGlass Pyrex® NMR Capillary Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Wilmad's capillary tubes are a low-cost solution for small volume NMR measurement.
- Available with both ends open or sealed at one end
- Tested at 400 MHz field
Made of ASTM Type 1 Class A glass.
Customers Who Bought This Also Bought