VWR® A-Series Balances, with NTEP Certificate
Supplier: VWR International
Certificates
About this item
VWR® A-Series balances incorporate state-of-the-art features for advanced weighing applications and documentation.
- Internal calibration
- Multiple weigh modes
- Voltage: 100 to 240 V
These economical balances offer a variety of standard features, such as internal calibration, GLP procedures, 17 weighing units, adjustable ambient condition settings, 11 application modes, and multilingual operation. The VWR® A-Series balances are the answer for organizations demanding precision and performance at an economical price.
Certifications: NTEP certified.
Specifications
- Catalog No:
- 76458-462
- 76458-464
- 76458-458
- 76458-460
- 76458-470
- 76458-472
- 76458-466
- 76458-468
- Operating temperature range:
- 10...40°C (50...104°F)
- 10...40°C (50...104°F)
- 10...40°C (50...104°F)
- 10...40°C (50...104°F)
- 10...40°C (50...104°F)
- 10...40°C (50...104°F)
- 10...40°C (50...104°F)
- 10...40°C (50...104°F)
- Communication interface:
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi