HM500 Series Half Mask Respirators, Honeywell Safety
Fournisseur: Honeywell Safety Products
Certificats
A propos de cet article
Offering breakthrough comfort, and reliable on the job safety, HM500 series half mask respirators are available in threaded/fixed or drop down styles.
- Soft material construction is comfortable on the skin to promote extended use
- Lighter than other masks with cartridges in the market
- Effective seal and good field of vision
- Easy cleaning and maintenance
- Low-profile cartridges enable a clear line of sight
- Drop down option allows for quick work breaks
- Compatible with all N-Series filters and cartridges
The Honeywell HM500 Series Half Mask is engineered as a lightweight, durable soft half mask with easy operation features. Available in non-drop down (threaded/fixed) and drop down versions, to ensure better comfort and convenience for extended wear. The drop-down option allows workers to remove the masks without taking off hard hats, earmuffs, or eyewear.
Fully compatible with the HM500, Honeywell N-Series filters and cartridges are quick and easy to attach, securely staying in place to provide confidence and safety on the job.
Customers Who Bought This Also Bought