A propos de cet article
LIBERO CE Accessory items below when paired with LIBERO CE data logger can be used to monitor dry shipper applications or temperature ranges.
- Used to monitor temperature range from −200 to +400 °C
This CFR 21 Part 11 compliant, reusable data logger is designed for ambient, cryo, dry ice, and liquid nitrogen shipment and storage monitoring. Covers a wide measurement range from −200 to +400 °C and can store as many as 75500 temperature values. Supports up to eight temperature and alarm zones, MKT, and duration as alarm criteria with an alarm function that can be turned on or off for cryo refill. No software is needed to generate a compliant CFR report because a raw data embedded PDF is patented by ELPRO and is designed not to lose or separate the raw data and PFD file. External PT100 probe is securely attached with M8 connector to enable reliable, superior temperature data transfer to the logger.