Dual Ended Teflon Coated Stainless Steel Lab Spatula, Eisco
# de catalogue 470328-212
Fournisseur: Wards
Spécifications
- Length:5.9"
- Description:Dual ended spatula
- Material:Teflon® coated
Spécifications
A propos de cet article
Features two identical flat spatula ends. Perfect for a variety of laboratory uses.
- Made of high quality stainless steel
- Coated in non-stick Teflon
- Extremely resistant to chemicals
- Autoclavable, reusable
- Feature one scoop end and one spatula end
Teflon coated stainless steel dual ended lab spatula features two identical flat spatula ends, 0.7" wide. Total length is 5.9" Great for adding small amounts of ingredients and mixing.
Customers Who Bought This Also Bought