Stainless Steel Lab Scoop with Spatula, Eisco
Fournisseur: Wards
Certificats
A propos de cet article
Features one scoop end and one spatula end with knife edge. Perfect for a variety of laboratory uses.
- Made of high quality polished stainless steel
- Autoclavable, reusable
- Feature one scoop end and one spatula end
Stainless steel lab scoop with spoon on one end and flat edge with on the other end. These scoops are great for adding small amounts of ingredients and mixing.
Customers Who Bought This Also Bought