Eisco Aluminium Dynamometers, Spring Balance
Fournisseur: Wards
Certificats
A propos de cet article
High resolution, durable aluminium spring balances are a great tool for accurately assessing the weight of an object in both grams and Newton's
- Premium design with aluminium tube
- High resolution, easy to read graduations
- Dual scale measures in grams and Newtons
- User friendly
- Useful for a variety of physics and physical science activities
Extremely durable with an aluminium tube, this dynamometer (spring balance) is easy to read, user firendly, and perfect for classroom use.
A dual scale allows measurements to be taken in grams (g) or Newtons (N).
Customers Who Bought This Also Bought