BioFit Elite Cleanroom ISO 7/ESD Swivel Chairs
Fournisseur: BioFit
Certificats
A propos de cet article
BioFit Elite Series seating, engineered for all-day comfort, is perfect for office and industrial workspaces. The ergonomic design enhances user comfort and productivity when sitting for prolonged periods.
- Easily adjusted controls to meet the needs of multiple shift users
- Mid-sized adjustable seat and back
- 5-legged wide aluminum base
- Dual-wheel nonmarring casters
BioFit seating solutions are crafted with user comfort, health and wellbeing in mind. Products meet or exceed rigorous industry standards and nearly all carry a 13-year warranty with a lifetime warranty on the pneumatic mechanisms.
The products are renowned for proven performance and reliability, backed by a history of over 75 years improving the workplace experience by providing ergonomic seating for laboratory, healthcare, technology, education, industrial, office and custom applications.
Certifications: LEED® compliant for use in commercial interiors; complies with ANSI/BIFMA X5.1 product safety and performance standards; meets standards for use under the purview of ANSI/ESD S20.20
Caution: WARNING: This product can expose you to chemicals including wood dust, which is known to the State of California to cause cancer. For more information go to www.p65warnings.ca.gov.
Spécifications
- Catalog No:
- 76449-994
- 76449-992
- 76449-998
- 76449-996
- 76449-986
- 76449-984
- 76449-990
- 76449-988
- 76450-018
- 76450-016
- 76450-022
- 76450-020
- 76450-010
- 76450-008
- 76450-014
- 76450-012
- 76449-980
- 76450-006
- 76450-004
- 76449-982
- 76450-026
- 76450-024
- 76450-002
- 76450-000
- Model:
- 76449-994
- 76449-992
- 76449-998
- 76449-996
- 76449-986
- 76449-984
- 76449-990
- 76449-988
- 76450-018
- 76450-016
- 76450-022
- 76450-020
- 76450-010
- 76450-008
- 76450-014
- 76450-012
- 76449-980
- 76450-006
- 76450-004
- 76449-982
- 76450-026
- 76450-024
- 76450-002
- 76450-000
- For use with:
- Worksurfaces 33 - 40" height
- Worksurfaces 30 - 34" height
- Worksurfaces 29 - 34" height
- Worksurfaces 37 - 44" height
- Worksurfaces 29 - 34" height
- Worksurfaces 37 - 44" height
- Worksurfaces 34 - 44" height
- Worksurfaces 31 - 38" height
- Worksurfaces 33 - 40" height
- Worksurfaces 30 - 34" height
- Worksurfaces 29 - 34" height
- Worksurfaces 37 - 44" height
- Worksurfaces 29 - 34" height
- Worksurfaces 37 - 44" height
- Worksurfaces 34 - 44" height
- Worksurfaces 31 - 38" height
- Worksurfaces 30 - 34" height
- Worksurfaces 33 - 40" height
- Worksurfaces 30 - 34" height
- Worksurfaces 33 - 40" height
- Worksurfaces 34 - 44" height
- Worksurfaces 31 - 38" height
- Worksurfaces 37 - 44" height
- Worksurfaces 31 - 38" height
- Frame colour/finish:
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Seat/back color:
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Blue
- Blue
- Blue
- Blue
- Blue
- Blue
- Blue
- Blue
- Black
- Blue
- Blue
- Black
- Blue
- Blue
- Black
- Black
- Seat/back material:
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Microcon vinyl
- Seat dimensions:
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- W18.5×D17×T3"
- Backrest dimensions:
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"
- W15.5×H14.5"