Cleanroom Facemasks, Gamma Irradiated, Green, Keystone Cleanroom Products
Fournisseur: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificats
A propos de cet article
These masks are with no-fog adhesive strip.
Certifications: Certificates of irradiation and conformance with each shipment.
Customers Who Bought This Also Bought