WHEATON® Single-Use Polyethylene Pasteur Pipettes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Ideal for general liquid transfer applications and extracting samples for analysis.
- Manufactured from soft, flexible, inert non-toxic polyethylene
- Ideal for safe and easy transfer of liquid samples
- Suitable for use as a freeze vial when the bulb is fully heat sealed
- Easy to handle with gloved hands
- Non-toxic material can be safely incinerated for disposal
- Available in easy-to-open, paper-plastic peel pouches
Available in a range of sizes from 1 ml to jumbo-sized pipettes with fine tip variants, there is a product to suit most user applications for use in areas such as blood banking, bacteriology, general life science and industry. The integral bulb design and single-use feature eliminate the need for plugging or autoclaving, helping to reduce the potential of cross-contamination during everyday sample handling.
Clear graduations on the 1 and 3 ml versions support ease of volume control. Jumbo pipettes are ideal for larger volume transfer or accessing liquids in taller vessels.
Customers Who Bought This Also Bought