Spécifications
- Description:Grids staining pad
- Cat. no.:76439-526
Spécifications
A propos de cet article
The grid staining pad is designed to simplify the preparation of cell samples for a transmission electron microscopy and make the process more time efficient. Ideal for ultrastructure studies of cells under a transmission electron microscope in neuroscience research.