Eisco Light Wall Glass Test Tubes
# de catalogue 470323-196
Fournisseur: Wards
Spécifications
- Capacity:15 ml
- Wall thickness:1.2 mm
- Description:Rimless tubes
- Cat. no.:470323-196
- O.D.×L:16×125 mm
Spécifications
A propos de cet article
Light wall rimless test tube for general use.
- Made from high quality Borosilicate 3.3 glass
- Feature fire polished, flat edge, and uniform wall thickness
- Autoclavable
- Superior chemical durability
- Low temperature gradient- can withstand direct flame, high temperatures and thermal shocks
Eisco Labs Borosilicate glass has the following heat tolerances: strain point –515 °C; annealing point 565 °C; softening point 820 °C. They manufacture their own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Customers Who Bought This Also Bought