Eisco Glass Weighing Bottles with Stopper, Low Form
Fournisseur: Wards
Certificats
A propos de cet article
Durable, high quality glass weighing bottle for a variety of laboratory uses.
- Borosilicate 3.3 glass
- Low form
- Autoclavable
- Superior chemical durability
- Low temperature gradient- can withstand high temperatures and thermal shocks
Eisco Labs Borosilicate glass has the following heat tolerances: strain point –515 °C; annealing point 565 °C and softening point 820 °C.
Delivery: Includes interchangable ground glass stopper.
Customers Who Bought This Also Bought