Nichipet Air Fully Autoclavable Micro Pipettes, Nichiryo America
Fournisseur: Nichiryo America
Certificats
A propos de cet article
Lightest weight, lighter stroke resistance, 8 different colors and easy calibration while maintaining higher accuracy and precision, and fully autoclaveble feature.
- Only 73.5 g (1000 µl)
- Easy handling and volume setting
- Easy self-calibration
- Longer second push (2 µl, 10 µl and 20 µl)
- Ergonomic shape
- Easy recognition (each volume sizes in different color)
- Filtered nozzles are used for large volume models (5000 µl and 10000 µl)
Available in eight different sizes covering a volume range of 0.1 µl to 10 ml, pipettors made of UV resistance material and lightest weight and lighter push resistance to reduce fatigue. A smooth, continuous, and adjustable micrometer replaces standard click-stop versions, allowing desired volumes to be dialed in for precise measurement. The 5000 µl, and 10 ml models are equipped with filtered nozzles.
Certifications: Test Report comply with ISO8655-6 standard is eclosed each pipette.
Caution: Rainin and LTS are a trademark of Mettler-Toledo, Rainin LLC.
Customers Who Bought This Also Bought