Intelsius® BioTherm (15...25 °C) Sample Transport Shippers, DGP Intelsius
Fournisseur: DGP INTELSIUS LLC
Certificats
A propos de cet article
Intelsius® BioTherm shippers are suitable for shipping temperature critical samples requiring a stable room temperature of between 15 and 25 °C.
- Passive cooling
- Compliant labels
- Reusable cold pack technology
- Regulatory compliant packaging
The BioTherm range of high performance 15 to 25 °C solutions are fully compliant to IATA, ADR, and DOT (49 CFR) transport regulations. These systems are qualified to ensure sample integrity for up to 96 hours against 15 to 25 °C temperature exposures. Each system is provided with handles and the correct compliance labeling to enable careful handling throughout the regulatory compliant cold chain. Cold packs are included.
Customers Who Bought This Also Bought