A propos de cet article
As a global supplier of compliant and certified sample transport packaging solutions, Intelsius® is committed to providing health professionals the products and tools they need to safely ship COVID-19 related samples and products.
- UN 3373 certified products
- COVID-19 sample compliant
- Variety of packaging types
Intelsius® has over 20 years of experience in delivering sample transport packaging solutions to a global market, with a range of products suitable for transporting UN 3373 Category B samples and temperature-sensitive payloads. These existing, proven solutions can help users transport samples, pharmaceuticals, and vaccines throughout the COVID-19 pandemic.
The demand for PathoPak, PathoShield, PathoSeal, and PathoPouch product ranges from health authorities across Europe, Asia, and the USA is expected to rise as testing and screening of populations increase. Designed for the transport of UN 3373 category B substances, these product ranges are fully compliant solutions for the transport of COVID-19 samples for testing and analysis.
PathoPak and Pathoshield products provide complete shipping solutions for UN 3373 sample transportation. PathoSeal and PathoPouch products provide 95kPa flexible secondaries which need to be paired with outers to compliantly transport samples for testing and analysis.