Intelsius® PathoShield Complete Sample Shipping Solution Packages, DGP Intelsius
Fournisseur: DGP INTELSIUS LLC
Certificats
A propos de cet article
The PathoShield range are complete shipping solutions designed to carry a wide range of primaries, including blood vials, swabs, and sample containers.
- Complete sample transport solution
- Includes 95kPa PathoPouch
- Pre-printed with required compliant markings
Each PathoShield complete system is pre-printed with the required markings and includes the relevant components to ship samples in compliance with transport regulations. The PathoShield solutions are ideal when transporting samples for COVID-19 testing and analysis.
Certifications: Complete Sample Shipping Solutions 3, 5, 6, and 7 are ISTA Certified Class 6.2, Category B. Complete Sample Shipping Solution 4 is ISTA Certified Class 6.2, Category A and B.
Customers Who Bought This Also Bought