A propos de cet article
This PLA filament is enhanced to improve the toughness of 3D builds.
- Diameter: 1.75 mm
- Spool size: 1 kg
- Print temperature: 210 to 220 °C
- Bed temperature: 65 °C
PLA, or Poly Lactic Acid, is a high-grade compostable bioplastic made from renewable natural resources. It's one of the most popular materials for 3D printing, as it has been perfected for ease of use with desktop 3D printers. PLA reduces calibration times, improves success rates, and ensures a consistent printing experience. With a minimal shrinkage factor, PLA filament will not warp or deform after cooling. PLA is recommended for use in schools and libraries. Other options, such as ABS filament, have certain benefits, but print with a noticeable plastic smell or give off potentially harmful chemicals. Using a PLA filament will keep the workspace safe and clean.
This filament from Inksmith is modified to retain the typical advantages of PLA, but with greatly improved toughness and reduced brittleness. Inksmith PLA+ will print directly on heated glass or a Cubicon bed, but will print best on heated PEI, Buildtak, or gluestick.