VWR® Precision Grip Gloves with Dots
Supplier: VWR International
Certificates
About this item
The VWR® precision grip glove with grip dots and polyurethane coating provide basic cut protection and enhanced grip. Ideal for handling, picking, packaging, automotive, or situations that require dexterity in a dry and/or dirty environment.
- Grip dots
- Nylon composition
- Nitrile (NBR) coated
- Gray knit glove with black coating
Certifications: Meets requirements for ANSI A1 Cut Protection (Level A). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications
- Catalog No:
- 76408-578
- 76408-576
- 76408-574
- 76408-572
- 76408-570
- 76408-568
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Nitrile coating
- Nitrile coating
- Nitrile coating
- Nitrile coating
- Nitrile coating
- Nitrile coating
- Gauge:
- 15
- 15
- 15
- 15
- 15
- 15
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile