VWR® Precision Grip Gloves, PU Coating
Supplier: VWR International
Certificates
About this item
The VWR® precision grip glove with polyurethane coating provide basic cut protection and grip. Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment.
- Nylon composition
- Polyurethane (PU) coated
- Black knit glove with black coating
Also great for package handling, picking, and packing.
Certifications: Meets requirements for ANSI A1 Cut Protection (Level A). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications
- Catalog No:
- 76408-560
- 76408-558
- 76408-556
- 76408-566
- 76408-564
- 76408-562
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Polyurethane coating
- Polyurethane coating
- Polyurethane coating
- Polyurethane coating
- Polyurethane coating
- Polyurethane coating
- Gauge:
- 15
- 15
- 15
- 15
- 15
- 15
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile