A propos de cet article
Strong, durable, and highly protective,the Intelsius® PathoPak range is UN certified for Category A shipments meeting IATA, ADR, and CFR 49 (DOT) transport regulations. The PathoPak ensures optimal protection for sample regardless of the specimen classification.
- Strong and durable
- Certified Class 6.2, Cat A and B
- Hard plastic
- Compliant labeling
The Intelsius® PathoPak range is a selection of complete sample transport solutions that are UN certified for the transportation of Category A (UN2814 and UN2900) and Category B (UN3373) infectious substances. Comprising of a 95kPa tested rigid secondary container, absorbent, bubble wrap, infectious labeling, and pre-printed outer with UN certification markings, the range is fully compliant meeting IATA, ADR, and CFR title 49 regulations for the transportation of infectious substances.
The PathoPak is ergonomically designed for fast and safe manual handling, meaning samples can be packaged and removed quickly as the PathoPak is perfect for large-scale eradication schemes and clinical trials. The PathoPak solutions are suitable for shipping a wide variety of primaries including blood tubes, specimen containers, swabs, blood collection/transfusion bags, and medical devices. It can also be used for transporting diagnostic samples and during screening programs.
Should any sample require delivery within a specific temperature range the PathoPak can be combined with an Intelsius® BioTherm. When combined with a BioTherm system sample integrity can be maintained for a minimum of 96 hours. Samples can be maintained between 2 to 8 °C, 15 to 25 °C, or at a frozen temperature using dry ice as coolant. The BioTherm systems are supplied with further compliance labeling and stable handles to ensure safe and compliant transportation throughout the cold chain.
The PathoPak system is designed to be used as a reusable system. The cardboard outer should be recycled and replaced after use with the secondary container sterilized and re-circulated. In certain cases, Intelsius® can offer a full sterilization and refurbishment service accepting the return of used secondaries, which will then be sterilised and recirculated with a new pre-printed cardboard outer, absorbents, and infectious labelling. This process enables a cost effective and environmentally sustainable approach for the transportation of biological and infectious substances. The refurbishment service also alleviates pressure on excessive stock holding requirements.