Intelsius® Pharmachill Reusable Cold Packs, DGP Intelsius
Supplier: DGP INTELSIUS LLC
Certificates
About this item
Pharmachill cold packs are reusable thermal media solutions using a water-based PCM ideal for maintaining payload temeprature at between 2 to 8 ˚C and 15 to 25 ˚C.
- Reusable
- Durable hard plastic
- Phase change material
Intelsius® produces a range of thermal media tailored to varied performance requirements. The Pharmachill range includes rigid, liquid-filled coolant bottles designed for use with temperature controlled packaging solutions.
Pharmachill Cold Packs feature a non-toxic, water based PCM. They have been specifically designed to be reusable, and therefore deliver a cost and environmental saving with each reuse.
Customers Who Bought This Also Bought