ContactUsLinkComponent
ImmuView Kit S. pneumoniae and L. pneumophila Urinary Antigen Test, Hardy Diagnostics
ImmuView Kit S. pneumoniae and L. pneumophila Urinary Antigen Test, Hardy Diagnostics
# de catalogue 76423-346
Fournisseur:  Hardy Diagnostics
Numéro CAS:  
Kit
ImmuView Kit S. pneumoniae and L. pneumophila Urinary Antigen Test, Hardy Diagnostics
# de catalogue 76423-346
Fournisseur:  Hardy Diagnostics
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Rapid ID test kit
  • Cat. no.:
    76423-346
Spécifications
A propos de cet article
ImmuView S. Pneumoniae and L. Pneumophila urinary antigen test kit, able to identify both S. Pneumoniae and Legionnaires disease at the same time-with just one test.
  • Quick
  • Accurate
  • Easy diagnosis
Quick, accurate and easy diagnosis is vital because S. pneumoniae is the leading cause of community acquired pneumonia. L. pneumophila causes costly hospital and ICU admissions. Not only that, the two diseases are difficult to tell apart, a significant challenge, as treatments for each disease are different.
Delivery: Each kit contains 1 positve and 1 negative control and 22 test strips.
Customers Who Bought This Also Bought