Compliant Labels, Intelsius®
Fournisseur: DGP INTELSIUS LLC
Certificats
A propos de cet article
Compliant labelling for infectious substances and dry ice. Other labels include orientation, security seals, exempt human specimen, and proper shipping labels.
- Compliant labels
- Wide assortment of UN regulation labels
- Easy to use
A comprehensive range of UN regulation labels to meet transport marking requirements. Category A and B labels feature a peel-off section with the required additional markings. Labels can be ordered in kit form, or in full reels. This is a cost effective solution for use with Intelsius outers suitable for Category A and B, and temperature controlled shipments.
Customers Who Bought This Also Bought