Tumbler Mixer 84 2T
# de catalogue 80700-570
Fournisseur: GOSSMANN PROJECTS INC. CA
Spécifications
- Description:Tumbler mixer 84 2T
- Size:W×D×H: 98×97×142 cm
- Cat. no.:80700-570
- Supplier No.:NO VPN
Spécifications
A propos de cet article
The versatile Tumbler Mixer is ideal for use in fire assay by mixing pulp samples with flux in fire assay crucibles. The mixer has two trays, each capable of holding up to 42×40 g crucibles. As an optional upgrade is an automatic timer as well as dual direction tumbling action to further enhance thorough mixing.
- Accommodates different crucible sizes ranging from 15 to 50g
- Provides thoroughly mixed samples within 8 to 15 minutes
- Adjustable rotational speed
A foam panel covered by plastic sheeting (sold separately) seals the crucibles so no additional crucible cover is required. The plastic sheeting can be easily replaced to avoid any cross contamination.
Delivery: Supplied complete with an exhaust port, which when attached to a dust collection system, eliminates any dust in the workplace making the working environment safer.
Customers Who Bought This Also Bought