Forced Convection Cleanroom Ovens, DES and DTS Series, Yamato
Fournisseur: Yamato Scientific
Certificats
A propos de cet article
These forced convection cleanroom ovens feature space saving, large volume class 100 clean oven.
- Improved visibility and operation of control panel
- Stable cleanliness through forced circulation with rear exhaust
- Display power consumption, CO₂ emissions and heater manipulated variables on the control panel
- Incorporate a maximum of 99 steps, 99 patterns program controller with repeat function
- Triac with zero-cross heater control
Offers several options such as recorder, manual/auto damper, N₂ gas introducer with flow meter and emergency switch.
DES830 convertible to high performance filter type maintaining Class 100 at stable and fluctuating temperature up to a maximum of 200 °C.
Safety features includes self-diagnostic functions (detection for temp. sensor failure, triac shortcircuit, automatic overheating prevention, heater line disconnect, main relay contact damage), earth leakage breaker, fan motor failure, key lock function, independent overheating prevention device.
Spécifications
- Catalog No:
- 76443-188
- 76443-186
- Insulation:
- Glass wool
- Glass wool
- Max. load per shelf:
- ~30 kg
- ~30 kg
- Sensor:
- K type Thermocouple dual sensor
- K type Thermocouple dual sensor