HALO® Guard Column Holder, Advanced Materials Technology
# de catalogue 76445-284
Fournisseur: Advanced Materials Technology
Spécifications
- Description:HALO® guard column holder, stainless steel
- Cat. no.:76445-284
Spécifications
A propos de cet article
Guard columm holder for HALO® guard column cartridges (required).
Customers Who Bought This Also Bought