HALO® C4, BIOCLASS, HPLC Columns, Advanced Materials Technology
Fournisseur: Advanced Materials Technology
Certificats
A propos de cet article
HALO® BIOCLASS C4 1000 Å Protein columns made with innovative large pore Fused-Core® particle technology provide best-in-class performance for high-resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO® 1000 Å columns are optimized for high performance in HPLC, UHPLC and LCMS large molecule applications, delivering excellent reproducibility and column lifetimes.
- Ideal for mAbs, ADCs and large proteins due to unrestricted access to the bonded phase
- Best-in-class resolution for full characterization of mAbs and their variants
- Exceptional mass transfer kinetics
- Outstanding low bleed performance and high temperature stability
Alternate selectivities with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
Spécifications
- Catalog No:
- 76443-392
- 76444-416
- 76444-488
- 76443-398
- 76444-422
- 76443-386
- 76444-410
- 76444-404
- 76446-076
- 76446-064
- 76444-482
- 76446-070
- 76444-476
- 76444-464
- 76444-392
- 76444-470
- 76444-398
- 76444-386
- 76446-058
- 76446-052
- 76444-458
- 76446-046
- 76444-452
- 76444-374
- 76444-446
- 76444-380
- 76537-466
- 76537-462
- 76537-458
- 76444-362
- 76444-440
- 76444-434
- 76444-358
- 76444-354
- 76444-506
- 76444-428
- 76444-342
- 76444-494
- 76444-500
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- USP code:
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26