HALO® Diphenyl, BIOCLASS, HPLC Columns, Advanced Materials Technology
Fournisseur: Advanced Materials Technology
Certificats
A propos de cet article
HALO® BIOCLASS Diphenyl 400 Å protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high-resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO® 400 Å columns are optimized for high performance in HPLC, UHPLC and LCMS large molecule applications, delivering excellent reproducibility and column lifetimes.
- Ideal for large protein analysis such as mAbs and polypeptides
- Well suited for high speed protein separations
- Exceptional recovery performance and temperature stability from 40 to 90 °C
- Outstanding low bleed performance
Alternate selectivities with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
Spécifications
- Catalog No:
- 76445-212
- 76445-150
- 76445-218
- 76445-140
- 76443-526
- 76445-156
- 76443-536
- 76445-202
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11