HALO® ES-C18, BIOCLASS, HPLC Columns, Advanced Materials Technology
Fournisseur: Advanced Materials Technology
Certificats
A propos de cet article
HALO® BIOCLASS ES-C18 400 Å Protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high-resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO 400 Å columns are optimized for high performance in HPLC, UHPLC and LCMS large molecule applications, delivering excellent reproducibility and column lifetimes.
- Ideal for large protein analysis such as mAbs and polypeptides
- Well suited for high speed protein separations
- Outstanding low bleed performance and high temperature stability
Alternate selectivity’s with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
Spécifications
- Catalog No:
- 76445-124
- 76445-214
- 76445-128
- 76445-204
- 76445-208
- 76445-120
- 76445-198
- 76445-112
- 76445-190
- 76445-104
- 76445-194
- 76445-186
- 76445-100
- 76445-178
- 76445-244
- 76445-092
- 76445-182
- 76445-170
- 76445-174
- 76445-166
- 76443-532
- 76445-162
- 76443-528
- 76445-158
- 76445-146
- 76445-236
- 76445-152
- 76445-240
- 76445-228
- 76445-232
- 76445-224
- 76443-522
- 76445-142
- 76445-132
- 76445-220
- 76445-136
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Temperature limit:
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1