HALO® ES-C18, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
Certificates
About this item
HALO® BIOCLASS ES-C18 1000 Å protein columns made with innovative large pore Fused-Core® particle technology provide best-in-class performance for high-resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns.
- Ideal for mAbs, ADCs and large proteins due to unrestricted access to the bonded phase
- Best-in-class resolution for full characterization of mAbs and their variants
- Exceptional mass transfer kinetics
- Outstanding low bleed performance and high temperature stability
HALO® 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Alternate selectivities with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
Specifications
- Catalog No:
- 76444-486
- 76444-372
- 76444-444
- 76443-390
- 76444-414
- 76444-492
- 76444-378
- 76443-396
- 76444-420
- 76443-384
- 76444-408
- 76446-074
- 76444-480
- 76446-068
- 76444-474
- 76444-352
- 76444-396
- 76444-438
- 76444-468
- 76444-390
- 76444-360
- 76444-402
- 76444-356
- 76444-504
- 76444-432
- 76444-384
- 76444-426
- 76446-062
- 76446-050
- 76446-056
- 76446-044
- 76444-462
- 76444-340
- 76444-450
- 76444-498
- 76444-456
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Temperature limit:
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1