Dionex Autosampler Probe Tip, Environmental Express
# de catalogue 76436-994
Fournisseur: Environmental Express
Spécifications
- Description:Environmental Express® stainless steel probe tip for Dionex AS/40 and AS-DV autosamplers
- Material:Stainless steel
- Cat. no.:76436-994
Spécifications
A propos de cet article
Optimize use of your Dionex Autosampler.
- Constructed with a durable stainless steel tip and FEP tubing
The stainless steel probe tip replaces the thermoplastic tip on the AS/40 and the AS-DV autosamplers. The steel tip will not scratch or deform ensuring an extremely long service life. The tip is fully rounded to provide a better engagement with the filter cap. Tip comes with 1 meter of 0.010×0.062" FEP tubing pre-inserted.
Customers Who Bought This Also Bought