AirLite™ Benchtop Enclosure, Environmental Express®
Fournisseur: Environmental Express
Certificats
A propos de cet article
Maintain clean air for more uniform digestions.
- Get HEPA-filtered air just about anywhere
- Exhaust can be connected to existing ductwork
- View digestions - clear PETG construction on all sides
- Easily access chamber - door opens a full 180°
- Cleanup is very easy with the removable, chemical-resistant bottom
- Quiet, easily accessible 117 cfm PVC coated fan
- HEPA filter can be changed in minutes
This enclosure is compatible with standard HotBlock® Digestion Systems.
Delivery: Systems include digestion cups and filtration assemblies. Tubing and vacuum source are not included.
Customers Who Bought This Also Bought