SimpleDist® Micro Tube Press, Environmental Express®
# de catalogue 76436-860
Fournisseur: Environmental Express
Spécifications
- Description:SimpleDist® Micro tube press
- Cat. no.:76436-860
- For Use With:C8000 SimpleDist® Micro distillation block
Spécifications
A propos de cet article
Obtain perfectly assembled tubes with this optional tube press.
- Save time and energy
Tubes fit into the press - Simply flip the lever for assembly.
Customers Who Bought This Also Bought