ContactUsLinkComponent
Accessories for FilterMate™ Digestion Cup Filters, Environmental Express
Accessories for FilterMate™ Digestion Cup Filters, Environmental Express
# de catalogue 76437-034
Fournisseur:  Environmental Express
Digestion Cup Holder, machined HDPE fixture adds stability when performing FilterMate filtrations, sample cup is placed into the center hole of the FilterMate holder prior to filtration, holder secures the digestion cup as the FilterMate is depressed through the liquid sample, Volume: 50 ml
Accessories for FilterMate™ Digestion Cup Filters, Environmental Express
# de catalogue 76437-034
Fournisseur:  Environmental Express
Spécifications
  • Description:
    FilterMate™ 50 ml digestion cup holder
  • Cat. no.:
    76437-034
Spécifications
A propos de cet article
Optimize the use of your FilterMate™ system with this digestion cup holder.
  • Constructed of HDPE
  • Compatible with 50 ml sample cups and FilterMate
This machined HDPE fixture adds stability when performing FilterMate filtrations. The 50 ml sample cup is placed into the center hole of the FilterMate holder prior to filtration. The holder secures the digestion cup as the FilterMate is depressed through the liquid sample. Works with 50 ml sample cup and FilterMate™.
Customers Who Bought This Also Bought