ProWeigh® and ProWeigh® Double Filters, Environmental Express®
Supplier: Environmental Express
Certificates
About this item
These ready-to-use filters for total and volatile suspended solids decrease sample prep time for your suspended solids testing.
- Approved for Standard Methods 2540
- Each filter is individually prepared, weighed, and labeled to be ready for use upon receipt
- Eliminate loose fibers and inaccurate weights with precise, laser-cut edges
- Quality control ensures filter integrity with visual inspection after each cutting, washing, and drying step
- Made from binderless borosilicate glass
All ProWeigh filters are prepared according to Standard Methods requirements. Filters are weighed, washed 3 times in deionized water, dried at 105 °C and weighed to 0.1 mg. Filters come packaged in a labeled aluminum pan with the weight of the filter printed on the label. In addition, the filter weight and ID number are printed in code 128 bar coding.
Delivery: Supplied with TKN100 base controller, 28-or 54-well TKN HotBlock, choice of 28 or 54× threaded (C6010) or non-threaded tubes (TKN6011), plus 28 or 54× teardrop stoppers (TKN6012), depending on options selected, see description below for full details.
Customers Who Bought This Also Bought