FilterMate™ Digestion Cup Filters, Environmental Express®
Fournisseur: Environmental Express
Certificats
A propos de cet article
Convenient filtration without a transfer step.
- Simple to use just press on plunger
- Filter 15 and 50 ml samples within sample cups
- No need for separate filtration devices before analyzing samples
Soil, sludge, wastewater, and challenging samples require filtration prior to analysis. FilterMate™ fits directly in the digestion vessel and is pushed through the liquid being filtered to the bottom of the cup with a detachable plunger. After filtration, the plunger is easily removed and discarded while the filter assembly remains in the bottom of the cup. Finished sample can be capped for storage or poured directly into an autosampler vial.
Ordering information: 100 filter assemblies and 100 plungers. Digestion cups sold separately.
Customers Who Bought This Also Bought