HemaPro Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Fournisseur: PROTIDE PHARMACEUTICALS
Certificats
A propos de cet article
Serum supplement for the culturing of human progenitor cells. Effective in studying cancer stem cells, iPSC, and mesenchymal stem cells.
- Defined, serum-free, xeno-free
- No hormones, antibiotics, growth factors, phenol red, Ig
- 50X concentrate
- Can be utilized with a wide variety of basal media
Certifications: cGMP manufactured.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.