Fluorescent Viewing Goggles, Noir medicals
# de catalogue 76420-054
Fournisseur: NOIR MEDICAL
Spécifications
- Description:Fluorescent viewing goggles
- Cat. no.:76420-054
Spécifications
A propos de cet article
Necessary protection when operating forensic light source.
Completely shield your eyes from UV A and UV B rays with these flexible orange viewing goggles. Clearly see latent prints and other evidence fluorescing with our wrap around goggles that meet ANSI standards and comfortably fit over prescription glasses.
Customers Who Bought This Also Bought